missing translation for 'onlineSavingsMsg'
Learn More

Lead(II) carbonate, basic, extra pure

Product Code. 10135503 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
500 g
2.5 kg
10 kg
Packaging:
Glass Bottle
Plastic bottle
Plastic drum
Unit Size:
10kg
2.5kg
500g
5g
This item is not returnable. View return policy

Product Code. 10135503

Brand: Thermo Scientific Acros 222710100

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1319-46-6 | C2H2O8Pb3 | 775.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1319-46-6
Molecular Formula C2H2O8Pb3
Molecular Weight (g/mol) 775.59
MDL Number MFCD00078155

Specifications

Melting Point 400.0°C
Color White
Physical Form Powder
Quantity 10 kg
Linear Formula (PbCO3)2·Pb(OH)2
Merck Index 15, 1002
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in acids
Formula Weight 775.59
Percent Purity ≥78% (Pb)
Grade Extra Pure
Packaging Plastic drum
Chemical Name or Material Lead(II) carbonate, basic, -325 Mesh
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Lead(II) carbonate, basic
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Reproductive toxicity Category 1A
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H360Df-May damage the unborn child. Suspected of damaging fertility.
  • H373-May cause damage to organs.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.