missing translation for 'onlineSavingsMsg'
Learn More

Lead(II) Carbonate Basic, Pure, SLR, Precipitated, Fisher Chemical™

£101.00

Chemical Identifiers

CAS 1319-46-6
Molecular Formula C2H2O8Pb3
MDL Number 78155
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10754951
View Documents
Fisher Chemical
L/1150/50
250 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Specifications

Chemical Identifiers

1319-46-6
C2H2O8Pb3
78155

Specifications

400°C
White
C2H2O8Pb3
775.59g/mol
Vapor Density: 9.21
1319-46-6
Powder/Solid
78155
Plastic powder jar
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Lead(II) carbonate, basic

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Reproductive toxicity Category 1A
  • Specific target organ toxicity after repeated exposure Category 2

Hazard Statement
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H360Df-May damage the unborn child. Suspected of damaging fertility.
  • H373-May cause damage to organs.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

SDS
Documents

Documents

Product Certifications