missing translation for 'onlineSavingsMsg'
Learn More
Lead(II) chloride, Reagent Grade, 99%, Thermo Scientific Chemicals Product Code.: 11381719

Lead(II) chloride, Reagent Grade, 99%, Thermo Scientific Chemicals

Product Code. 11381719
500 g
Click to view available options
Quantity:
50 g
500 g
2 kg
This item is not returnable. View return policy

Product Code. 11381719

Brand: Thermo Scientific Alfa Aesar 012349.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Analytical Reagent, Preparation of Lead Salts, As Solder and Flux

It is employed in the manufacture of infrared transmitting glass, and ornamental glass called arena glass. It also finds application as analytical reagent, in preparation of lead salts, as solder and flux, for provision of lead titanate, and used in the purification process of ores.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7758-95-4
Molecular Formula Cl2Pb
Molecular Weight (g/mol) 278.10
MDL Number MFCD00011157
InChI Key HWSZZLVAJGOAAY-UHFFFAOYSA-L
Synonym lead dichloride, lead ii chloride, lead chloride, plumbous chloride, lead 2+ chloride, lead chloride pbcl2, pbcl2, ccris 7565, lead ii chloride, ultra dryShow MoreShow Less
PubChem CID 24459
IUPAC Name dichlorolead
SMILES [Cl-].[Cl-].[Pb++]

Specifications

Boiling Point 951°C
Melting Point 501°C
Color White
Physical Form Crystalline Powder
Quantity 500 g
Assay Percent Range 99%
Solubility Information Soluble in hot water,alkali hydroxides and NH4Cl solution. Insoluble in cold water and alcohol.
Formula Weight 278.1
Grade Reagent
Odor Odorless
Density 5.85
Chemical Name or Material Lead(II) chloride
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberU16H082 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number012349.18,012349.A3,012349.36
Lot NumberP17A020 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number012349.18,012349.A3,012349.36
Lot NumberZ18D004 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number012349.18,012349.A3,012349.36
Lot NumberC14N32 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number012349.18,012349.A3,012349.36
Lot NumberK29S055 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number012349.18,012349.A3,012349.36

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Lead(II) chloride, Reagent Grade, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.