missing translation for 'onlineSavingsMsg'
Learn More
Lead(II) cyclohexanebutyrate, 94%, Thermo Scientific™ Product Code.: 11493507

Lead(II) cyclohexanebutyrate, 94%, Thermo Scientific™

Product Code. 11493507
100 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 11493507

Brand: Thermo Scientific Alfa Aesar A17280.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62637-99-4
Molecular Formula C20H34O4Pb
Molecular Weight (g/mol) 545.688
MDL Number MFCD00036403
InChI Key CCJZHCZMNGUOCA-UHFFFAOYSA-L
Synonym lead cyclohexanebutyrate, lead bis 4-cyclohexylbutyrate, cyclohexanebutanoic acid, lead 2+ salt, cyclohexanebutanoic acid, lead 2+ salt 2:1, lead 2+ bis 4-cyclohexylbutanoate, acmc-20akns, lead ii cyclohexanebutyrate, 4-cyclohexylbutanoate; lead 2+, biscyclohexanebutyric acid lead ii salt, $l^ 2-lead 2+ ion bis 4-cyclohexylbutanoateShow MoreShow Less
PubChem CID 112896
IUPAC Name 4-cyclohexylbutanoate;lead(2+)
SMILES C1CCC(CC1)CCCC(=O)[O-].C1CCC(CC1)CCCC(=O)[O-].[Pb+2]

Specifications

Quantity 100 g
UN Number UN2291
Formula Weight 545.68
Percent Purity 94%
Chemical Name or Material Lead(II) cyclohexanebutyrate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberC3552B Certificate TypeCertificate of Analysis Date06/12/2021 Catalog Number
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Lead(II) cyclohexanebutyrate, 94%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.