missing translation for 'onlineSavingsMsg'
Learn More

Lead(II) formate, 90+%, Thermo Scientific™

Product Code. 11446927
Click to view available options
Quantity:
50 g
250 g
1000 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 811-54-1
Molecular Formula C2H2O4Pb
Molecular Weight (g/mol) 297.234
MDL Number MFCD00050834
InChI Key ZGVSCHRFRACEIO-UHFFFAOYSA-L
Synonym lead diformate, lead formate, formic acid, lead 2+ salt, unii-2kjp65ymf4, lead ii formate, lead 2+ formate, 2kjp65ymf4, lead formate pb o2ch 2, lead formate pb hco2 2, formic acid, lead 2+ salt 2:1
PubChem CID 61216
IUPAC Name lead(2+);diformate
SMILES C(=O)[O-].C(=O)[O-].[Pb+2]

Specifications

Melting Point 190°C (decomposition)
Quantity 50 g
UN Number UN2291
Merck Index 14,5409
Formula Weight 297.23
Percent Purity ≥90%
Chemical Name or Material Lead(II) formate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'