missing translation for 'onlineSavingsMsg'
Learn More

Lead(II) hexafluorosilicate dihydrate, Thermo Scientific™

Product Code. 11368108
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1310-03-8
Molecular Formula F6H4O20Pb9Si6
Molecular Weight (g/mol) 2471.312
MDL Number MFCD00049509
InChI Key UCXBPZOKVHNQLY-UHFFFAOYSA-N
Synonym acmc-20aknf, fluoro trioxido silane;lead 2+ ;dihydrate, silicate 2-,hexafluoro-, lead 2+ 1:1 , dihydrate 9ci
PubChem CID 57347846
IUPAC Name fluoro(trioxido)silane;lead(2+);dihydrate
SMILES O.O.[O-][Si]([O-])([O-])F.[O-][Si]([O-])([O-])F.[O-][Si]([O-])([O-])F.[O-][Si]([O-])([O-])F.[O-][Si]([O-])([O-])F.[O-][Si]([O-])([O-])F.[Pb+2].[Pb+2].[Pb+2].[Pb+2].[Pb+2].[Pb+2].[Pb+2].[Pb+2].[Pb+2]

Specifications

Physical Form Powder
Quantity 50 g
Linear Formula PbSiF6·2H2O
UN Number UN2291
Formula Weight 385.26 (349.26 Anhydrous)
Odor Odorless
Chemical Name or Material Lead(II) hexafluorosilicate dihydrate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.