missing translation for 'onlineSavingsMsg'
Learn More

Lead(II,IV) oxide, red, 98%

Product Code. 10781301 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
100 g
500 g
5 kg
Packaging:
Glass bottle
Unit Size:
100g
500g
5g
5kg
This item is not returnable. View return policy

Product Code. 10781301

Brand: Thermo Scientific Acros 221110050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 1314-41-6 | O4Pb3 | 685.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1314-41-6
Molecular Formula O4Pb3
Molecular Weight (g/mol) 685.57
MDL Number MFCD00078491
InChI Key XMFOQHDPRMAJNU-UHFFFAOYSA-N
Synonym lead tetroxide, orange lead, lead oxide, red, lead ii,iv oxide, po4b3, lead oxide, red, lr, lead oxide, red powder, 1-2 mum, lead oxide, red, saj first grade, lead oxide, powder trace metals basis
PubChem CID 16685188
IUPAC Name 1,3,5,7-tetraoxa-2$l^{2},4,6$l^{2}-triplumbaspiro[3.3]heptane
SMILES O1[Pb]O[Pb]12O[Pb]O2

Specifications

Melting Point 500.0°C
Color Orange
Physical Form Powder
Quantity 5 g
Assay Percent Range 97.5% min. (PB3O4)
Linear Formula 2PbO·PbO2
Merck Index 15,548
Solubility Information Solubility in water: insoluble. Other solubilities: partly soluble in acids, insoluble in alcohol
Formula Weight 685.57
Percent Purity 98%
Packaging Glass bottle
Chemical Name or Material Lead(II, IV) oxide, red, 98%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.