Learn More
Lead(II) nitrate, 99%, for analysis, Thermo Scientific Chemicals
Chemical Identifiers
CAS | 10099-74-8 |
---|---|
Molecular Formula | N2O6Pb |
Molecular Weight (g/mol) | 331.20 |
MDL Number | MFCD00011153 |
InChI Key | RLJMLMKIBZAXJO-UHFFFAOYSA-N |
Synonym | lead dinitrate, lead nitrate, lead ii nitrate, plumbous nitrate, lead 2+ nitrate, lead nitrate pb no3 2, nitrate de plomb french, lead ii nitrate 1:2, nitric acid, lead 2+ salt |
PubChem CID | 24924 |
ChEBI | CHEBI:37187 |
SMILES | [Pb++].[O-][N+]([O-])=O.[O-][N+]([O-])=O |
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
10099-74-8 | |
331.20 | |
RLJMLMKIBZAXJO-UHFFFAOYSA-N | |
24924 | |
[Pb++].[O-][N+]([O-])=O.[O-][N+]([O-])=O |
N2O6Pb | |
MFCD00011153 | |
lead dinitrate, lead nitrate, lead ii nitrate, plumbous nitrate, lead 2+ nitrate, lead nitrate pb no3 2, nitrate de plomb french, lead ii nitrate 1:2, nitric acid, lead 2+ salt | |
CHEBI:37187 |
Specifications
470.0°C | |
98.5% min. (Complexometry) | |
Pb(NO3)2 | |
15,5469 | |
Solubility in water: 343g/L. | |
[Pb++].[O-][N+]([O-])=O.[O-][N+]([O-])=O | |
331.20 | |
CHEBI:37187 | |
99% | |
4.53 | |
Lead(II) nitrate |
10099-74-8 | |
N2O6Pb | |
MFCD00011153 | |
lead dinitrate, lead nitrate, lead ii nitrate, plumbous nitrate, lead 2+ nitrate, lead nitrate pb no3 2, nitrate de plomb french, lead ii nitrate 1:2, nitric acid, lead 2+ salt | |
RLJMLMKIBZAXJO-UHFFFAOYSA-N | |
lead(2+);dinitrate | |
24924 | |
331.2 | |
Analytical | |
4.5300g/mL |
Safety and Handling
GHS H Statement
Harmful if inhaled.
May damage the unborn child.
Suspected of damaging fertility.
May cause damage to organs through prolonged or repeated exposure.
Very toxic to aquatic life with long lasting effects.
GHS P Statement
Use personal protective equipment as required.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Do not breathe dust/fume
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 233-245-9
- Lead(II) nitrate
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Serious eye damage/eye irritation Category 1
- Oxidising solid Category 2
- Reproductive toxicity Category 1A
- Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
- H318-Causes serious eye damage.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- H360Df-May damage the unborn child. Suspected of damaging fertility.
- H373-May cause damage to organs.
- H302-Harmful if swallowed.
- H332-Harmful if inhaled.
- H272-May intensify fire; oxidiser.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
RUO – Research Use Only