missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lead(II) Nitrate, 99+%, Technical, Fisher Chemical
Click to view available options
Quantity:
500 g
Packaging:
Plastic powder jar
Specifications
Melting Point | 470°C |
CAS | 10099-74-8 |
Color | White |
pH | 3 to 4 |
Physical Form | Solid |
Quantity | 500 g |
Molecular Formula | N2O6Pb |
MDL Number | MFCD00011153 |
Synonym | lead dinitrate, lead nitrate, lead ii nitrate, plumbous nitrate, lead 2+ nitrate, lead nitrate pb no3 2, nitrate de plomb french, lead ii nitrate 1:2, nitric acid, lead 2+ salt |
InChI Key | RLJMLMKIBZAXJO-UHFFFAOYSA-N |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'