Learn More
Lead(II) nitrate, 99%, Thermo Scientific Chemicals
Description
Lead(II) nitrate is employed in industrial applications such as heat stabilization in nylon and polyesters and coatings of photothermographic paper. It is utilized to improve the leaching process in the gold cyanidation. It finds applications as a lead paint as well as a source of dinitrogen tetroxide. As an oxidant, it is used in the oxidation of benzylic halides to aldehydes in organic synthesis. It is also used in the preparation of dithiocarbamates from isothiocyanates. It acts as a scavenger of bromide in nucleophilic substitution reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lead(II) nitrate is employed in industrial applications such as heat stabilization in nylon and polyesters and coatings of photothermographic paper. It is utilized to improve the leaching process in the gold cyanidation. It finds applications as a lead paint as well as a source of dinitrogen tetroxide. As an oxidant, it is used in the oxidation of benzylic halides to aldehydes in organic synthesis. It is also used in the preparation of dithiocarbamates from isothiocyanates. It acts as a scavenger of bromide in nucleophilic substitution reactions.
Solubility
Soluble in water. Insoluble in nitric acid. Sparingly soluble in methanol.
Notes
Incompatible with strong reducing agents, organic materials and powdered metals.

Specifications
Specifications
Melting Point | 470°C |
Quantity | 100 g |
UN Number | UN1469 |
Merck Index | 14,5414 |
Solubility Information | Soluble in water. Insoluble in nitric acid. Sparingly soluble in methanol. |
Formula Weight | 331.2 |
Percent Purity | 99% |
Odor | Odorless |
Density | 4.53 |
Chemical Name or Material | Lead(II) nitrate |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
M29L028 | Certificate of Analysis | 03/02/2025 | A16345.0E,A16345.36,A16345.22 |
X09K056 | Certificate of Analysis | 30/10/2024 | A16345.0E,A16345.36,A16345.22 |
10252880 | Certificate of Analysis | 28/10/2024 | A16345.0E,A16345.36,A16345.22 |
T25K045 | Certificate of Analysis | 20/08/2024 | A16345.0E,A16345.36,A16345.22 |
P06K016 | Certificate of Analysis | 11/06/2024 | A16345.0E,A16345.36,A16345.22 |
Safety and Handling





- Lead(II) nitrate
- Danger
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Serious eye damage/eye irritation Category 1
- Oxidising solid Category 2
- Reproductive toxicity Category 1A
- Specific target organ toxicity after repeated exposure Category 2
- H272-May intensify fire; oxidiser.
- H302-Harmful if swallowed.
- H318-Causes serious eye damage.
- H332-Harmful if inhaled.
- H360Df-May damage the unborn child. Suspected of damaging fertility.
- H373-May cause damage to organs.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- MIXTURE LIST-Contains : Lead(II) nitrate
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.