missing translation for 'onlineSavingsMsg'
Learn More

Lead(Ii) Nitrate, ACS Reagent, Honeywell Fluka™

ACS Reagent,≥99.0%

£40.89 - £222.52

Chemical Identifiers

CAS 10099-74-8
Molecular Formula N2O6Pb
Molecular Weight (g/mol) 331.20
MDL Number MFCD00011153
InChI Key RLJMLMKIBZAXJO-UHFFFAOYSA-N
Synonym lead dinitrate, lead nitrate, lead ii nitrate, plumbous nitrate, lead 2+ nitrate, lead nitrate pb no3 2, nitrate de plomb french, lead ii nitrate 1:2, nitric acid, lead 2+ salt
PubChem CID 24924
ChEBI CHEBI:37187
IUPAC Name lead(2+);dinitrate
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15612270
View Documents Promotion Details
Honeywell-Fluka
228621-100G
100 g
£40.89
100g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15632270
View Documents Promotion Details
Honeywell-Fluka
228621-500G
500 g
£60.91
500g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15622270
View Documents Promotion Details
Honeywell-Fluka
228621-2.5KG
2.5 kg
£222.52
2.50kg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications

Chemical Identifiers

10099-74-8
331.20
RLJMLMKIBZAXJO-UHFFFAOYSA-N
24924
lead(2+);dinitrate
N2O6Pb
MFCD00011153
lead dinitrate, lead nitrate, lead ii nitrate, plumbous nitrate, lead 2+ nitrate, lead nitrate pb no3 2, nitrate de plomb french, lead ii nitrate 1:2, nitric acid, lead 2+ salt
CHEBI:37187

Specifications

10099-74-8
Pb(NO3)2
1469
RLJMLMKIBZAXJO-UHFFFAOYSA-N
lead(2+);dinitrate
24924
331.21g/mol
ACS Reagent
N2O6Pb
MFCD00011153
lead dinitrate, lead nitrate, lead ii nitrate, plumbous nitrate, lead 2+ nitrate, lead nitrate pb no3 2, nitrate de plomb french, lead ii nitrate 1:2, nitric acid, lead 2+ salt
[Pb++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
331.20
CHEBI:37187
≥99%
Lead(Ii) Nitrate
Videos
Safety and Handling

Safety and Handling

P201-P210-P220-P280-P305 + P351 + P338 + P310-P308 + P313

H272-H302 + H332-H318-H360Df-H373-H410

missing translation for 'einecsNumber' : 233-245-9

missing translation for 'rtecsNumber' : OG2100000

Product Identifier
  • Lead(II) nitrate

Signal Word
  • Danger

Hazard Category
  • Oxidising solid Category 2
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity after repeated exposure Category 2
  • Reproductive toxicity Category 1A
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1

Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H302 + H332-Harmful if swallowed or if inhaled.
  • H318-Causes serious eye damage.
  • H360Df-May damage the unborn child. Suspected of damaging fertility.
  • H373-May cause damage to organs.
  • H410-Very toxic to aquatic life with long lasting effects.

Precautionary Statement
  • P201-Obtain special instructions before use.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

SDS
Documents

Documents

Product Certifications