missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lead(II) Nitrate, Certified AR for Analysis, Fisher Chemical™
£366.00
Chemical Identifiers
CAS | 10099-74-8 |
---|---|
Molecular Formula | N2O6Pb |
Molecular Weight (g/mol) | 331.20 |
MDL Number | MFCD00011153 |
InChI Key | RLJMLMKIBZAXJO-UHFFFAOYSA-N |
Synonym | lead dinitrate, lead nitrate, lead ii nitrate, plumbous nitrate, lead 2+ nitrate, lead nitrate pb no3 2, nitrate de plomb french, lead ii nitrate 1:2, nitric acid, lead 2+ salt |
PubChem CID | 24924 |
ChEBI | CHEBI:37187 |
SMILES | [Pb++].[O-][N+]([O-])=O.[O-][N+]([O-])=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10080230
|
Fisher Chemical
L/1500/60 |
1 kg | N/A | ||||||
Description
Chemical Identifiers
10099-74-8 | |
331.20 | |
RLJMLMKIBZAXJO-UHFFFAOYSA-N | |
24924 | |
[Pb++].[O-][N+]([O-])=O.[O-][N+]([O-])=O |
N2O6Pb | |
MFCD00011153 | |
lead dinitrate, lead nitrate, lead ii nitrate, plumbous nitrate, lead 2+ nitrate, lead nitrate pb no3 2, nitrate de plomb french, lead ii nitrate 1:2, nitric acid, lead 2+ salt | |
CHEBI:37187 |
Specifications
470°C | |
White | |
Solid | |
MFCD00011153 | |
RLJMLMKIBZAXJO-UHFFFAOYSA-N | |
lead(2+);dinitrate | |
24924 | |
331.2g/mol |
10099-74-8 | |
3 to 4 | |
N2O6Pb | |
lead dinitrate, lead nitrate, lead ii nitrate, plumbous nitrate, lead 2+ nitrate, lead nitrate pb no3 2, nitrate de plomb french, lead ii nitrate 1:2, nitric acid, lead 2+ salt | |
[Pb++].[O-][N+]([O-])=O.[O-][N+]([O-])=O | |
331.20 | |
CHEBI:37187 | |
Plastic powder jar |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Lead(II) nitrate
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Serious eye damage/eye irritation Category 1
- Oxidising solid Category 2
- Reproductive toxicity Category 1A
- Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
- H318-Causes serious eye damage.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- H360Df-May damage the unborn child. Suspected of damaging fertility.
- H373-May cause damage to organs.
- H302-Harmful if swallowed.
- H332-Harmful if inhaled.
- H272-May intensify fire; oxidiser.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P281-Use personal protective equipment as required.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ A§ if you feel unwell.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title