missing translation for 'onlineSavingsMsg'
Learn More
Lead(II) nitrate, White powder, Puratronic™, 99.999% (Metals basis), Thermo Scientific Chemicals Product Code.: 11376797

Lead(II) nitrate, White powder, Puratronic™, 99.999% (Metals basis), Thermo Scientific Chemicals

Product Code. 11376797
1 kg
Click to view available options
Quantity:
50 g
250 g
1 kg
This item is not returnable. View return policy

Product Code. 11376797

Brand: Thermo Scientific Alfa Aesar 010727.A1

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Employed in industrial applications such as heat stabilization in nylon and polyesters, coatings of photothermographic paper, gold cyanidation and pyrotechnics. Finds application in supramolecular chemistry. Strong oxidizing agent, and an alternative to the Sommelet reaction for the oxidation of benzylic halides to aldehydes. Used in the preparation of isothiocyanates from dithiocarbamates. Scavenger of bromide in nucleophilic substitution reactions. Manufacture of dinitrogen tetroxide in indurtrial scale.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10099-74-8
Molecular Formula N2O6Pb
Molecular Weight (g/mol) 331.20
MDL Number MFCD00011153
InChI Key RLJMLMKIBZAXJO-UHFFFAOYSA-N
Synonym lead dinitrate, lead nitrate, lead ii nitrate, plumbous nitrate, lead 2+ nitrate, lead nitrate pb no3 2, nitrate de plomb french, lead ii nitrate 1:2, nitric acid, lead 2+ salt
PubChem CID 24924
ChEBI CHEBI:37187
SMILES [Pb++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Melting Point 470°C (decomposition)
Color White
Physical Form Powder
Quantity 1 kg
Assay Percent Range 99.999% (Metals basis)
Solubility Information soluble in water. Insoluble in nitric acid. Sparingly soluble in methanol.
IUPAC Name λ²-lead(2+) dinitrate
Formula Weight 331.2
Odor Odorless
Sensitivity Hygroscopic
Density 4.53
Chemical Name or Material Lead(II) nitrate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberX27J036 Certificate TypeCertificate of Analysis Date03/11/2023 Catalog Number010727.30,010727.18,010727.A1

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Lead(II) nitrate, White powder, Puratronic™, 99.999% (Metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.