missing translation for 'onlineSavingsMsg'
Learn More

Lead(II) nitrate, White powder, Puratronic™, 99.999% (Metals basis), Thermo Scientific Chemicals

Catalog Number 11396797
Click to view available options
Quantity:
50 g
250 g
1 kg
This item is not returnable. View return policy
10099-74-8
N2O6Pb
331.20
MFCD00011153
RLJMLMKIBZAXJO-UHFFFAOYSA-N
lead dinitrate, lead nitrate, lead ii nitrate, plumbous nitrate, lead 2+ nitrate, lead nitrate pb no3 2, nitrate de plomb french, lead ii nitrate 1:2, nitric acid, lead 2+ salt
24924
CHEBI:37187
[Pb++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
This item is not returnable. View return policy
10099-74-8
N2O6Pb
331.20
MFCD00011153
RLJMLMKIBZAXJO-UHFFFAOYSA-N
lead dinitrate, lead nitrate, lead ii nitrate, plumbous nitrate, lead 2+ nitrate, lead nitrate pb no3 2, nitrate de plomb french, lead ii nitrate 1:2, nitric acid, lead 2+ salt
24924
CHEBI:37187
[Pb++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

Employed in industrial applications such as heat stabilization in nylon and polyesters, coatings of photothermographic paper, gold cyanidation and pyrotechnics. Finds application in supramolecular chemistry. Strong oxidizing agent, and an alternative to the Sommelet reaction for the oxidation of benzylic halides to aldehydes. Used in the preparation of isothiocyanates from dithiocarbamates. Scavenger of bromide in nucleophilic substitution reactions. Manufacture of dinitrogen tetroxide in indurtrial scale.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 470°C (decomposition)
Color White
Physical Form Powder
Quantity 250 g
Assay Percent Range 99.999% (Metals basis)
Solubility Information soluble in water. Insoluble in nitric acid. Sparingly soluble in methanol.
IUPAC Name lead(2+);dinitrate
Formula Weight 331.2
Odor Odorless
Sensitivity Hygroscopic
Density 4.53
Chemical Name or Material Lead(II) nitrate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'