Learn More
Lead(II) oxide, 99.9%, Thermo Scientific Chemicals
£38.59 - £65.02
Chemical Identifiers
CAS | 1317-36-8 |
---|---|
Molecular Formula | OPb |
Molecular Weight (g/mol) | 223.20 |
MDL Number | MFCD00011164 |
InChI Key | YEXPOXQUZXUXJW-UHFFFAOYSA-N |
Synonym | lead monoxide, lead ii oxide, lead oxide pbo, massicot, massicotite, lead monooxide, lead protoxide, plumbous oxide, litharge pure, lead oxide yellow |
PubChem CID | 14827 |
IUPAC Name | oxolead |
SMILES | O=[Pb] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11306638
|
Thermo Scientific Alfa Aesar
012220.36 |
500 g |
£38.59
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11316638
|
Thermo Scientific Alfa Aesar
012220.A3 |
2 kg |
£65.02
2kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
It is employed mostly in lead-based industrial glass and industrial ceramics, including computer components. Lead monoxide is used as an intermediate/precursor in the manufacture of several products, for example water proof cements, lubricants, lubricating oils, inorganic pigments, lead soaps, petroleum refining, rubber, cathode ray tube glass, and polyvinyl chloride (PVC). It is useful for lead acid batteries as cathode and anode. Lead monoxide scores significant applications in oil, gas and chemical manufactures. It is an efficient catalyst for condensation reactions in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1317-36-8 | |
223.20 | |
YEXPOXQUZXUXJW-UHFFFAOYSA-N | |
14827 | |
O=[Pb] |
OPb | |
MFCD00011164 | |
lead monoxide, lead ii oxide, lead oxide pbo, massicot, massicotite, lead monooxide, lead protoxide, plumbous oxide, litharge pure, lead oxide yellow | |
oxolead |
Specifications
886°C | |
Yellow | |
99.9% | |
MFCD00011164 | |
Soluble in concentrated alkali,hydrochloric acid,and ammonium chloride. Insoluble in water,dilute alkali and alcohol. | |
O=[Pb] | |
223.20 | |
223.19 | |
9.53 g/cm3 at 20°C |
1317-36-8 | |
Powder | |
OPb | |
lead monoxide, lead ii oxide, lead oxide pbo, massicot, massicotite, lead monooxide, lead protoxide, plumbous oxide, litharge pure, lead oxide yellow | |
YEXPOXQUZXUXJW-UHFFFAOYSA-N | |
oxolead | |
14827 | |
Odorless | |
Lead(II) oxide |
Safety and Handling
Avoid exposure - obtain special instructions before use. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste. Avoid release to the environment. Refer to special instructions/Safety data sheets.
P201-P202-P260-P264b-P270-P271-P281-P301+P312-P304+P340-P308+P313-P330-P501c
H302+H332-H350-H360Df-H372
missing translation for 'dotInformation' : DOT Class: 9, Packaging Group: III
missing translation for 'einecsNumber' : 215-267-0
missing translation for 'rtecsNumber' : OG1750000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Lead(II) oxide
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Reproductive toxicity Category 1A
- Specific target organ toxicity after repeated exposure Category 2
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
- Carcinogenicity Category 2
- Reproductive toxicity on or via lactation
Hazard Statement
- H302-Harmful if swallowed.
- H332-Harmful if inhaled.
- H360Df-May damage the unborn child. Suspected of damaging fertility.
- H373-May cause damage to organs.
- H410-Very toxic to aquatic life with long lasting effects.
- H351-Suspected of causing cancer.
- H362-May cause harm to breast-fed children.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P405-Store locked up.
- P501-Dispose of contents/container to .
RUO – Research Use Only