missing translation for 'onlineSavingsMsg'
Learn More

Lead(II) oxide, 99.9+%, (trace metal basis), <10 microns, powder

Product Code. 10063570 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Unit Size:
100g
2.5kg
500g
This item is not returnable. View return policy

Product Code. 10063570

Brand: Thermo Scientific Acros 315855000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1317-36-8 | OPb | 223.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

For ultra trace analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1317-36-8
Molecular Formula OPb
Molecular Weight (g/mol) 223.20
MDL Number MFCD00011164
InChI Key YEXPOXQUZXUXJW-UHFFFAOYSA-N
Synonym lead monoxide, lead ii oxide, lead oxide pbo, massicot, massicotite, lead monooxide, lead protoxide, plumbous oxide, litharge pure, lead oxide yellow
PubChem CID 14827
SMILES O=[Pb]

Specifications

Boiling Point 1470.0°C
Melting Point 886.0°C
Color Yellow
Physical Form Powder
Quantity 500 g
Assay Percent Range 99.9+%
Linear Formula PbO
Fieser 01,536; 02,233
Merck Index 15, 5468
Solubility Information Solubility in water: 70.2 mg/l (20°C).
IUPAC Name oxolead
Formula Weight 223.19
Percent Purity ≥99.9%
Grade Trace Metal Basis
Packaging Glass bottle
Chemical Name or Material Lead(II) oxide, <10 micron, 99.9+%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.