Learn More
Lead(II) selenide, 99.999+% (metals basis), Thermo Scientific Chemicals
£270.35 - £766.82
Chemical Identifiers
CAS | 12069-00-0 |
---|---|
Molecular Formula | PbSe |
Molecular Weight (g/mol) | 286.20 |
MDL Number | MFCD00016273 |
InChI Key | GGYFMLJDMAMTAB-UHFFFAOYSA-N |
Synonym | lead selenide, lead ii selenide, lead ii selenide trace metals basis 25g |
PubChem CID | 61550 |
IUPAC Name | selanylidenelead |
SMILES | [Se]=[Pb] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15442067
|
Thermo Scientific Alfa Aesar
044916.14 |
25 g |
£270.35
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15452067
|
Thermo Scientific Alfa Aesar
044916.22 |
100 g |
£766.82
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Lead(II) selenide is used in the production of infrared detectors for thermal imaging. It is also used as quantum dots in nanocrystal solar cell. It is an important material and finds applications in photographic plates, and photo resistors and optoelectronic devices.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
12069-00-0 | |
286.20 | |
GGYFMLJDMAMTAB-UHFFFAOYSA-N | |
61550 | |
[Se]=[Pb] |
PbSe | |
MFCD00016273 | |
lead selenide, lead ii selenide, lead ii selenide trace metals basis 25g | |
selanylidenelead |
Specifications
12069-00-0 | |
Fused Lump | |
PbSe | |
lead selenide, lead ii selenide, lead ii selenide trace metals basis 25g | |
GGYFMLJDMAMTAB-UHFFFAOYSA-N | |
selanylidenelead | |
61550 | |
Lead(II) selenide |
Gray | |
>99.999% (Metals basis) | |
MFCD00016273 | |
Insoluble in water. | |
[Se]=[Pb] | |
286.20 | |
286.15 |
Safety and Handling
P201-P202-P260-P264b-P270-P271-P281-P301+P310-P304+P340-P308+P313-P311-P330-P501c
H301+H331-H350-H360Df-H373
missing translation for 'dotInformation' : DOT Class: 9, Packaging Group: III
missing translation for 'einecsNumber' : 235-109-4
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Lead(II) selenide
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Acute toxicity Category 2
- Reproductive toxicity Category 1A
- Specific target organ toxicity after repeated exposure Category 2
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
- H301-Toxic if swallowed.
- H330-Fatal if inhaled.
- H360-May damage fertility or the unborn child.
- H373-May cause damage to organs.
- H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P320-Specific treatment is urgent (see on this label).
- P405-Store locked up.
- P501-Dispose of contents/container to .
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.