missing translation for 'onlineSavingsMsg'
Learn More
Lead(II) sulfate, 99%, Thermo Scientific Chemicals Product Code.: 10686923

Lead(II) sulfate, 99%, Thermo Scientific Chemicals

Product Code. 10686923
100 g, Plastic bottle
Click to view available options
Quantity:
100 g
500 g
Packaging:
Plastic bottle
This item is not returnable. View return policy

Product Code. 10686923

Brand: Thermo Scientific Acros 387271000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7446-14-2
Molecular Formula O4PbS
Molecular Weight (g/mol) 303.25
MDL Number MFCD00011166
InChI Key KEQXNNJHMWSZHK-UHFFFAOYSA-L
Synonym lead ii sulfate, lead sulfate, anglislite, lead sulphate, mulhouse white, lead bottoms, lead monosulfate, fast white, milk white, natural anglesiteShow MoreShow Less
PubChem CID 24008
IUPAC Name lead(2+);sulfate
SMILES [O-]S(=O)(=O)[O-].[Pb+2]

Specifications

Melting Point 1170.0°C
Color White
Physical Form Crystalline Powder
Quantity 100 g
Assay Percent Range 99%
Merck Index 15, 5475
Solubility Information Solubility in water: slightly soluble
Formula Weight 303.25
Percent Purity 99%
Identification Confirmed
Packaging Plastic bottle
Chemical Name or Material Lead(II) sulfate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0459303 Certificate TypeCertificate of Analysis Date11/01/2024 Catalog Number387271000,387275000
Lot NumberA0456268 Certificate TypeCertificate of Analysis Date11/09/2023 Catalog Number387271000,387275000
Lot NumberA0456269 Certificate TypeCertificate of Analysis Date11/09/2023 Catalog Number387271000,387275000
Lot NumberA0450831 Certificate TypeCertificate of Analysis Date02/05/2023 Catalog Number387271000,387275000
Lot NumberA0392259 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number387271000,387275000
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Lead(II) sulfate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Reproductive toxicity Category 1A
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H360Df-May damage the unborn child. Suspected of damaging fertility.
  • H373-May cause damage to organs.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Lead(II) sulfate, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.