missing translation for 'onlineSavingsMsg'
Learn More

Lead(II) telluride, 99.999% (metals basis)

Product Code. 11329239
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11329239

Brand: Thermo Scientific Alfa Aesar 088285.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1314-91-6 | PbTe | 334.80 g/mol

Lead(II) telluride is used to make thermoelectric converter (cooler, generator, heat), infrared detectors and tunnel diode. It is also an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1314-91-6
Molecular Formula PbTe
Molecular Weight (g/mol) 334.80
MDL Number MFCD00016274
InChI Key OCGWQDWYSQAFTO-UHFFFAOYSA-N
Synonym lead telluride, lead ii telluride, unii-v1og6oa4bj, v1og6oa4bj, plumbanetellone, bleitellurid, lead monotelluride, lead ii telluride trace metals basis
PubChem CID 4389803
IUPAC Name tellanylidenelead
SMILES [Te]=[Pb]

Specifications

Melting Point 917°C
Physical Form Pieces
Quantity 100 g
UN Number UN3284
Merck Index 14,5422
Solubility Information Insoluble in water.
Formula Weight 334.79
Percent Purity 99.999%
Odor Product specific
Density 8.164 g/mL
Assay (metals basis)
Chemical Name or Material Lead (II) telluride
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.