Learn More
Lead(II) tetrafluoroborate, 50% w/w aq. soln., Thermo Scientific Chemicals
Used in Tin-Lead Electroplating
£130.65 - £284.42
Chemical Identifiers
CAS | 13814-96-5 |
---|---|
Molecular Formula | B2F8Pb |
Molecular Weight (g/mol) | 380.80 |
MDL Number | MFCD00016270 |
InChI Key | UQGXAENNKXXYNF-UHFFFAOYSA-N |
Synonym | lead ii tetrafluoroborate, lead fluoroborate, lead ii tetrafluoroborate solution, lead boron fluoride, lead tetrafluoroborate, unii-wax1e0vj84, lead ii fluoroborate, acmc-1bpsn, wax1e0vj84, lead 2+ ditetrafluoroborate |
PubChem CID | 12598180 |
SMILES | [Pb++].F[B-](F)(F)F.F[B-](F)(F)F |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11334947
|
Thermo Scientific Alfa Aesar
041956.A1 |
1 kg |
£130.65
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11344947
|
Thermo Scientific Alfa Aesar
041956.D5 |
3 x 1 kg |
£284.42
3kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Used in tin-lead electroplatingLead(II) tetrafluoroborate is used as a catalyst involved in calixarene formation through hydrogen-bonded dimmers and in the preparation of linear polyesters. It is involved in the development and optimization of a bipolar lead-boron tetrafluoride accumulator. It is an electroplating solution for coating metal objects with lead and curing agent for epoxy resins. Further, it is used in printing lines, tin-lead alloy plating, lead soldering and also as a circuit board tin-lead alloy plating.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
13814-96-5 | |
380.80 | |
UQGXAENNKXXYNF-UHFFFAOYSA-N | |
12598180 |
B2F8Pb | |
MFCD00016270 | |
lead ii tetrafluoroborate, lead fluoroborate, lead ii tetrafluoroborate solution, lead boron fluoride, lead tetrafluoroborate, unii-wax1e0vj84, lead ii fluoroborate, acmc-1bpsn, wax1e0vj84, lead 2+ ditetrafluoroborate | |
[Pb++].F[B-](F)(F)F.F[B-](F)(F)F |
Specifications
13814-96-5 | |
Liquid | |
MFCD00016270 | |
Miscible with water. | |
[Pb++].F[B-](F)(F)F.F[B-](F)(F)F | |
380.80 | |
380.81 | |
1.62 |
Colorless | |
B2F8Pb | |
lead ii tetrafluoroborate, lead fluoroborate, lead ii tetrafluoroborate solution, lead boron fluoride, lead tetrafluoroborate, unii-wax1e0vj84, lead ii fluoroborate, acmc-1bpsn, wax1e0vj84, lead 2+ ditetrafluoroborate | |
UQGXAENNKXXYNF-UHFFFAOYSA-N | |
lead(2+);ditetrafluoroborate | |
12598180 | |
Odorless | |
Lead(II) tetrafluoroborate |
Safety and Handling
P201-P202-P260-P264b-P270-P271-P281-P301+P312-P304+P340-P308+P313-P330-P501c
H302+H332-H350-H360Df-H373
missing translation for 'dotInformation' : DOT Class: 8, Packaging Group: III
missing translation for 'einecsNumber' : 237-486-0
missing translation for 'rtecsNumber' : ED2700000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Lead(II) tetrafluoroborate, 50% w/w aqueous solution
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Reproductive toxicity Category 1A
- Specific target organ toxicity after repeated exposure Category 2
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
- H302-Harmful if swallowed.
- H360Df-May damage the unborn child. Suspected of damaging fertility.
- H373-May cause damage to organs.
- H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P273-Avoid release to the environment.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P405-Store locked up.
- P501-Dispose of contents/container to .
Supplemental information
- MIXTURE LIST-Contains : Lead(II) tetrafluoroborate
RUO – Research Use Only