missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lead(II) titanium oxide, 99.5%, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
500 g
2 kg
Description
Lead(II) titanium oxide is widely used in ceramic capacitor, ceramic filters, PTC thermal resistor, pressure sensitive resistor and the performance of transducer field.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Color | Yellow |
Physical Form | Powder |
Quantity | 2 kg |
UN Number | UN3288 |
Solubility Information | Insoluble in water. |
Formula Weight | 303.09 |
Percent Purity | 99.5% |
Odor | Odorless |
Density | 7.520 g/mL |
Chemical Name or Material | Lead(II) titanium oxide |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'