missing translation for 'onlineSavingsMsg'
Learn More

Lead(II) titanium oxide, 99.5%, Thermo Scientific Chemicals

Product Code. 11397737
Click to view available options
Quantity:
100 g
500 g
2 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

Lead(II) titanium oxide is widely used in ceramic capacitor, ceramic filters, PTC thermal resistor, pressure sensitive resistor and the performance of transducer field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12060-00-3
Molecular Formula O3PbTi
Molecular Weight (g/mol) 303.064
MDL Number MFCD00011159
InChI Key NKZSPGSOXYXWQA-UHFFFAOYSA-N
Synonym lead titanate, lead titanium trioxide, lead ii titanate, lead titanium oxide, lead titanium oxide pbtio3, titanic acid, lead salt, titanox l, titanicacid,leadsalt, o3ti.pb, lead titanate, powder,
PubChem CID 16211560
IUPAC Name dioxido(oxo)titanium;lead(2+)
SMILES [O-][Ti](=O)[O-].[Pb+2]

Specifications

Color Yellow
Physical Form Powder
Quantity 500 g
UN Number UN3288
Solubility Information Insoluble in water.
Formula Weight 303.09
Percent Purity 99.5%
Odor Odorless
Density 7.520 g/mL
Chemical Name or Material Lead(II) titanium oxide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'