Learn More
Lead(II) trifluoroacetate hemihydrate, Thermo Scientific Chemicals
£115.00 - £332.00
Chemical Identifiers
CAS | 4146-73-0 |
---|---|
Molecular Formula | C4F6O4Pb |
Molecular Weight (g/mol) | 433.23 |
MDL Number | MFCD00061594 |
InChI Key | XVULNTYAIWEMSW-UHFFFAOYSA-L |
Synonym | lead trifluoroacetate, lead ii 2,2,2-trifluoroacetate, acmc-1asut, bis trifluoroacetoxy lead ii |
PubChem CID | 71311182 |
IUPAC Name | lead(2+);2,2,2-trifluoroacetate |
SMILES | C(=O)(C(F)(F)F)[O-].C(=O)(C(F)(F)F)[O-].[Pb+2] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11343377
|
Thermo Scientific Alfa Aesar
017553.09 |
10 g |
£115.00
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11353377
|
Thermo Scientific Alfa Aesar
017553.18 |
50 g | |||||||
Description
It is employed in pyrolytic deposition of fluoride glass.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
4146-73-0 | |
433.23 | |
XVULNTYAIWEMSW-UHFFFAOYSA-L | |
71311182 | |
C(=O)(C(F)(F)F)[O-].C(=O)(C(F)(F)F)[O-].[Pb+2] |
C4F6O4Pb | |
MFCD00061594 | |
lead trifluoroacetate, lead ii 2,2,2-trifluoroacetate, acmc-1asut, bis trifluoroacetoxy lead ii | |
lead(2+);2,2,2-trifluoroacetate |
Specifications
4146-73-0 | |
White | |
C4F6O4Pb | |
MFCD00061594 | |
lead trifluoroacetate, lead ii 2,2,2-trifluoroacetate, acmc-1asut, bis trifluoroacetoxy lead ii | |
XVULNTYAIWEMSW-UHFFFAOYSA-L | |
lead(2+);2,2,2-trifluoroacetate | |
71311182 | |
Powder |
147°C to 153°C | |
Odorless | |
Pb(CF3COO)2·0.5H2O | |
UN2291 | |
Soluble in water,trifluoroacetic acid,tetrahydrofuran,acetone. | |
C(=O)(C(F)(F)F)[O-].C(=O)(C(F)(F)F)[O-].[Pb+2] | |
433.23 | |
442.23(433.22 Anhydrous) | |
Lead(II) trifluoroacetate hemihydrate |
Safety and Handling
GHS H Statement
H360-H373-H302-H332
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.
P201-P202-P260-P264b-P270-P271-P281-P301+P312-P304+P340-P308+P313-P330-P501c
H302+H332-H360Df-H373
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: LEAD COMPOUNDS, SOLUBLE, N.O.S.
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
- Lead(II) trifluoroacetate hemihydrate
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Reproductive toxicity Category 1A
- Specific target organ toxicity after repeated exposure Category 2
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
- H302-Harmful if swallowed.
- H332-Harmful if inhaled.
- H360Df-May damage the unborn child. Suspected of damaging fertility.
- H373-May cause damage to organs.
- H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
- P201-Obtain special instructions before use.
- P273-Avoid release to the environment.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P501-Dispose of contents/container to .
RUO – Research Use Only