Learn More
Lead(II) zirconium oxide, 99.7% (metals basis excluding Hf), Hf 1% max, Thermo Scientific Chemicals
£132.66 - £637.17
Chemical Identifiers
CAS | 12060-01-4 |
---|---|
Molecular Formula | O3PbZr |
Molecular Weight (g/mol) | 346.421 |
MDL Number | MFCD00016275 |
InChI Key | RSCSNKLWUCUPDE-UHFFFAOYSA-N |
Synonym | lead zirconate, o3zr.pb, dioxido oxo zirconium;lead 2+, $l^ 2-lead 2+ ion oxozirconiumbis olate |
PubChem CID | 16212542 |
IUPAC Name | dioxido(oxo)zirconium;lead(2+) |
SMILES | [O-][Zr](=O)[O-].[Pb+2] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11356988
|
Thermo Scientific Alfa Aesar
014682.22 |
100 g |
£132.66
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11366988
|
Thermo Scientific Alfa Aesar
014682.36 |
500 g |
£637.17
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
It is practically applied in the area of electroceramics.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
12060-01-4 | |
346.421 | |
RSCSNKLWUCUPDE-UHFFFAOYSA-N | |
16212542 | |
[O-][Zr](=O)[O-].[Pb+2] |
O3PbZr | |
MFCD00016275 | |
lead zirconate, o3zr.pb, dioxido oxo zirconium;lead 2+, $l^ 2-lead 2+ ion oxozirconiumbis olate | |
dioxido(oxo)zirconium;lead(2+) |
Specifications
12060-01-4 | |
O3PbZr | |
UN3077 | |
Insoluble in all solvents. | |
[O-][Zr](=O)[O-].[Pb+2] | |
346.421 | |
346.42 | |
Odorless | |
(metals basis excluding Hf), Hf 1% max. |
Powder | |
MFCD00016275 | |
lead zirconate, o3zr.pb, dioxido oxo zirconium;lead 2+, $l^ 2-lead 2+ ion oxozirconiumbis olate | |
RSCSNKLWUCUPDE-UHFFFAOYSA-N | |
dioxido(oxo)zirconium;lead(2+) | |
16212542 | |
99.7% | |
7.0 g/mL | |
Lead(II) zirconium oxide |
Safety and Handling
GHS H Statement
H360-H373-H302-H332
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.
P201-P202-P260-P264b-P270-P271-P281-P301+P312-P304+P340-P308+P313-P330-P501c
H302+H332-H350-H360Df-H373
missing translation for 'dotInformation' : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
missing translation for 'einecsNumber' : 235-039-4
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Lead zirconium oxide
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Reproductive toxicity Category 1A
- Specific target organ toxicity after repeated exposure Category 2
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
- H302-Harmful if swallowed.
- H332-Harmful if inhaled.
- H360Df-May damage the unborn child. Suspected of damaging fertility.
- H373-May cause damage to organs.
- H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P405-Store locked up.
- P501-Dispose of contents/container to .
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.