missing translation for 'onlineSavingsMsg'
Learn More
Lead(II) zirconium oxide, Puratronic™, 99.997% (metals basis), Thermo Scientific Chemicals Product Code.: 11375047

Lead(II) zirconium oxide, Puratronic™, 99.997% (metals basis), Thermo Scientific Chemicals

Product Code. 11375047
50 g
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy

Product Code. 11375047

Brand: Thermo Scientific Alfa Aesar 040754.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12060-01-4
Molecular Formula O3PbZr
Molecular Weight (g/mol) 346.421
MDL Number MFCD00016275
InChI Key RSCSNKLWUCUPDE-UHFFFAOYSA-N
Synonym lead zirconate, o3zr.pb, dioxido oxo zirconium;lead 2+, $l^ 2-lead 2+ ion oxozirconiumbis olate
PubChem CID 16212542
IUPAC Name dioxido(oxo)zirconium;lead(2+)
SMILES [O-][Zr](=O)[O-].[Pb+2]

Specifications

Physical Form Powder
Quantity 50 g
UN Number UN3077
Formula Weight 346.42
Percent Purity 99.997%
Grade Puratronic™
Odor Odorless
Density 7.0 g/mL
Assay (metals basis)
Chemical Name or Material Lead(II) zirconium oxide

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

3 results found
Lot Number Certificate Type Date Catalog Number
Lot Number91356 Certificate TypeCertificate of Analysis Date24/11/2021 Catalog Number040754.18,040795.TD,040795.TC,040795.TB,040754.09,040754.30
Lot NumberS91356 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog Number040754.18,040795.TD,040795.TC,040795.TB,040754.09,040754.30
Lot NumberB23U023 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog Number040754.18,040795.TD,040795.TC,040795.TB,040754.09,040754.30

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Lead(II) zirconium oxide, Puratronic™, 99.997% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.