missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lead(Iv) Dioxide, Extra Pure, SLR, Fisher Chemical™
Click to view available options
Quantity:
500 g
Specifications
Specifications
Melting Point | 290°C |
Color | Black |
pH | 6 to 7 |
Physical Form | Powder/Solid |
Quantity | 500 g |
IUPAC Name | plumbanedione |
Formula Weight | 239.19g/mol |
Packaging | Plastic powder jar |
Safety and Handling
Product Identifier
- Lead(IV) oxide
- Danger
- Oxidising solid Category 3
- Acute toxicity Category 4
- Reproductive toxicity Category 1A
- Specific target organ toxicity after repeated exposure Category 2
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
- H272-May intensify fire; oxidiser.
- H302-Harmful if swallowed.
- H332-Harmful if inhaled.
- H373-May cause damage to organs.
- H360Df-May damage the unborn child. Suspected of damaging fertility.
- H410-Very toxic to aquatic life with long lasting effects.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P201-Obtain special instructions before use.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'