missing translation for 'onlineSavingsMsg'
Learn More

Lead(Iv) Dioxide, Extra Pure, SLR, Fisher Chemical™

Product Code. p-7073263 Shop All Fisher Chemical Products
Click to view available options
Quantity:
500 g
Unit Size:
500g
This item is not returnable. View return policy

Product Code. 10796481

Brand: Fisher Chemical L/1350/53

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 1309-60-0
Molecular Formula O2Pb
Molecular Weight (g/mol) 239.20
MDL Number MFCD00011165
InChI Key YADSGOSSYOOKMP-UHFFFAOYSA-N
Synonym lead dioxide, lead iv oxide, lead peroxide, lead superoxide, lead oxide pbo2, lead brown, lead oxide brown, lead peroxide pbo2, bioxyde de plomb french
PubChem CID 14793
SMILES O=[Pb]=O

Specifications

Melting Point 290°C
Color Black
pH 6 to 7
Physical Form Powder/Solid
Quantity 500 g
IUPAC Name plumbanedione
Formula Weight 239.19g/mol
Packaging Plastic powder jar
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Lead(IV) oxide
Signal Word
  • Danger
Hazard Category
  • Oxidising solid Category 3
  • Acute toxicity Category 4
  • Reproductive toxicity Category 1A
  • Specific target organ toxicity after repeated exposure Category 2
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H373-May cause damage to organs.
  • H360Df-May damage the unborn child. Suspected of damaging fertility.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P201-Obtain special instructions before use.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.