missing translation for 'onlineSavingsMsg'
Learn More
Lead(IV) oxide, ACS, 97.0% min, Thermo Scientific Chemicals Product Code.: 11331829

Lead(IV) oxide, ACS, 97.0% min, Thermo Scientific Chemicals

Product Code. 11331829
100 g
Click to view available options
Quantity:
100 g
500 g
2 kg
This item is not returnable. View return policy

Product Code. 11331829

Brand: Thermo Scientific Alfa Aesar 012483.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For Use in Battery Electrodes, Manufacturing of Pigments and Dyes, As Analytical Lab Reagent

It finds applications as an electrode in lead acid batteries, as anode in electroplating copper and zinc in sulfate solution, production of glyoxylic acid, manufacturing of pigments and dyes, high voltage lightening arrestors, and as an analytical lab reagent. Lead dioxide is a potent oxidizing agent which is employed in the manufacture of matches, pyrotechnics and other chemicals. It can be used in the curing of sulfide-based polymers. It oxidizes Mn(II) into Mn(VII), and Cr(III) into Cr(VI).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Maximum level of impurities: Acid-insoluble matter 0.2%, Carbon compounds (as C) 0.04%, Cl 0.002%, NO3 0.02%, SO4 0.05%, Mn 5ppm, Ca 0.02%, Cu 0.05%, K 0.05%, Na 0.1%
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1309-60-0
Molecular Formula O2Pb
Molecular Weight (g/mol) 239.20
MDL Number MFCD00011165
InChI Key YADSGOSSYOOKMP-UHFFFAOYSA-N
Synonym lead dioxide, lead iv oxide, lead peroxide, lead superoxide, lead oxide pbo2, lead brown, lead oxide brown, lead peroxide pbo2, bioxyde de plomb french
PubChem CID 14793
SMILES O=[Pb]=O

Specifications

Melting Point 290°C (decomposition)
Color Brown to Black
Physical Form Powder
Quantity 100 g
Assay Percent Range 97.0% min.
Solubility Information Insoluble in water and alcohol. Soluble in acetic acid.
IUPAC Name plumbanedione
Formula Weight 239.19
Grade ACS
Odor Odorless
Density 9.38
Chemical Name or Material Lead(IV) oxide
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberP24L020 Certificate TypeCertificate of Analysis Date26/03/2025 Catalog Number012483.36,012483.A3,012483.22
Lot NumberZ23H037 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number012483.36,012483.22
Lot NumberM31G017 Certificate TypeCertificate of Analysis Date28/12/2021 Catalog Number012483.36,012483.22
Lot NumberU06H013 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number012483.36,012483.22
Lot NumberK07E09 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number012483.36,012483.22
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Lead(IV) oxide
Signal Word
  • Danger
Hazard Category
  • Oxidising solid Category 2
  • Acute toxicity Category 4
  • Reproductive toxicity Category 1A
  • Specific target organ toxicity after repeated exposure Category 2
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H360Df-May damage the unborn child. Suspected of damaging fertility.
  • H373-May cause damage to organs.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P201-Obtain special instructions before use.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Lead(IV) oxide, ACS, 97.0% min, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.