Learn More
Lead plate, 6.35mm (0.25in) thick, 99.9% (metals basis), Thermo Scientific Chemicals
£45.02 - £133.67
Chemical Identifiers
CAS | 7439-92-1 |
---|---|
Molecular Formula | Pb |
Molecular Weight (g/mol) | 207.20 |
MDL Number | MFCD00134050 |
InChI Key | WABPQHHGFIMREM-UHFFFAOYSA-N |
Synonym | plumbum, metal, blei, lead, elemental, rough bullion, plomo, glover, element, flake, omaha & grant |
PubChem CID | 5352425 |
ChEBI | CHEBI:27889 |
IUPAC Name | lead |
SMILES | [Pb] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11317447
|
Thermo Scientific Alfa Aesar
012603.VA |
15 x 15 cm |
£45.02
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11307447
|
Thermo Scientific Alfa Aesar
012603.RF |
30 x 30 cm |
£133.67
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7439-92-1 | |
207.20 | |
WABPQHHGFIMREM-UHFFFAOYSA-N | |
5352425 | |
lead |
Pb | |
MFCD00134050 | |
plumbum, metal, blei, lead, elemental, rough bullion, plomo, glover, element, flake, omaha & grant | |
CHEBI:27889 | |
[Pb] |
Specifications
Gray | |
Pb | |
plumbum, metal, blei, lead, elemental, rough bullion, plomo, glover, element, flake, omaha & grant | |
[Pb] | |
207.20 | |
CHEBI:27889 | |
Odorless | |
Plate | |
Lead plate, 6.35mm (0.25 in.) thick |
7439-92-1 | |
MFCD00134050 | |
WABPQHHGFIMREM-UHFFFAOYSA-N | |
lead | |
5352425 | |
99.9% | |
≈1.6kg/15x15cm | |
(metals basis) |
Safety and Handling
GHS H Statement
H360-H373-H302-H332
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.
P201-P202-P260-P263-P264b-P270-P281-P308+P313-P501c
H350-H360FD-H362-H372
missing translation for 'einecsNumber' : 231-100-4
missing translation for 'rtecsNumber' : OF7525000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only