missing translation for 'onlineSavingsMsg'
Learn More
Lead powder, -100 mesh, 99.95% (metals basis), Thermo Scientific Chemicals Product Code.: 15432407

Lead powder, -100 mesh, 99.95% (metals basis), Thermo Scientific Chemicals

Product Code. 15432407
1 kg
Click to view available options
Quantity:
250 g
1 kg
This item is not returnable. View return policy

Product Code. 15432407

Brand: Thermo Scientific Alfa Aesar 045602.A1

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7439-92-1
Molecular Formula Pb
Molecular Weight (g/mol) 207.20
MDL Number MFCD00134050
InChI Key WABPQHHGFIMREM-UHFFFAOYSA-N
Synonym plumbum, metal, blei, lead, elemental, rough bullion, plomo, glover, element, flake, omaha & grant
PubChem CID 5352425
ChEBI CHEBI:27889
IUPAC Name lead
SMILES [Pb]

Specifications

Color Black to Gray
Appearance Gray to black atomized powder
Assay Percent Range 99.95% (Metals basis)
Quantity 1 kg
Odor Odorless
Form Powder
Chemical Name or Material Lead

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberZ17H005 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number045604.09,045602.30,045602.A1
Lot NumberY09H020 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number045604.09,045602.30,045602.A1
Lot NumberN26B036 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number045604.09,045602.30,045602.A1
Lot NumberI14Y059 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number045604.09,045602.30,045602.A1
Lot NumberP04A030 Certificate TypeCertificate of Analysis Date24/11/2021 Catalog Number045604.09,045602.30,045602.A1

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Lead powder, -100 mesh, 99.95% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.