Learn More
Lead powder, -200 mesh, 99.9% (metals basis), Thermo Scientific Chemicals
£44.32 - £312.56
Chemical Identifiers
CAS | 7439-92-1 |
---|---|
Molecular Formula | Pb |
Molecular Weight (g/mol) | 207.20 |
MDL Number | MFCD00134050 |
InChI Key | WABPQHHGFIMREM-UHFFFAOYSA-N |
Synonym | plumbum, metal, blei, lead, elemental, rough bullion, plomo, glover, element, flake, omaha & grant |
PubChem CID | 5352425 |
ChEBI | CHEBI:27889 |
IUPAC Name | lead |
SMILES | [Pb] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11391278
|
Thermo Scientific Alfa Aesar
010223.30 |
250 g |
£44.32
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11381278
|
Thermo Scientific Alfa Aesar
010223.A1 |
1 kg |
£67.23
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11371278
|
Thermo Scientific Alfa Aesar
010223.A7 |
5 kg |
£312.56
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7439-92-1 | |
207.20 | |
WABPQHHGFIMREM-UHFFFAOYSA-N | |
5352425 | |
lead |
Pb | |
MFCD00134050 | |
plumbum, metal, blei, lead, elemental, rough bullion, plomo, glover, element, flake, omaha & grantShow More | |
CHEBI:27889 | |
[Pb] |
Specifications
Black to Gray | |
Gray to black powder | |
Pb | |
plumbum, metal, blei, lead, elemental, rough bullion, plomo, glover, element, flake, omaha & grant | |
[Pb] | |
207.20 | |
CHEBI:27889 | |
Odorless | |
Lead |
7439-92-1 | |
99.9% (Metals basis) | |
MFCD00134050 | |
WABPQHHGFIMREM-UHFFFAOYSA-N | |
lead | |
5352425 | |
-200 Mesh | |
Powder |
Safety and Handling
P201-P202-P260-P263-P264b-P270-P281-P308+P313-P501c
H350-H360FD-H362-H372
missing translation for 'dotInformation' : DOT Class: 9, Packaging Group: III
missing translation for 'einecsNumber' : 231-100-4
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Lead powder
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Reproductive toxicity Category 1A
- Specific target organ toxicity after repeated exposure Category 2
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
- H302-Harmful if swallowed.
- H332-Harmful if inhaled.
- H360Df-May damage the unborn child. Suspected of damaging fertility.
- H373-May cause damage to organs.
- H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P405-Store locked up.
- P501-Dispose of contents/container to .
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.