missing translation for 'onlineSavingsMsg'
Learn More

Lead rod, 12.7mm (0.5in) dia, Puratronic™, 99.9998% (metals basis)

Product Code. 11341119
Click to view available options
Quantity:
50 g
210 g
4 x 210 g
Unit Size:
210g
50g
840g
This item is not returnable. View return policy

Product Code. 11341119

Brand: Thermo Scientific Alfa Aesar 011346.97

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7439-92-1 | Pb | 207.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7439-92-1
Molecular Formula Pb
Molecular Weight (g/mol) 207.20
MDL Number MFCD00134050
InChI Key WABPQHHGFIMREM-UHFFFAOYSA-N
Synonym plumbum, metal, blei, lead, elemental, rough bullion, plomo, glover, element, flake, omaha & grant
PubChem CID 5352425
ChEBI CHEBI:27889
IUPAC Name lead
SMILES [Pb]

Specifications

Color Gray
Quantity 210 g
Odor Odorless
Weight ≈3.5cm/50g
Form Rod
Percent Purity 99.9998%
Grade Puratronic
Assay (metals basis)
Chemical Name or Material Lead rod, 12.7mm (0.5 in.) dia., May be in multiple pieces

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.