Learn More
Lead standard solution, for AAS, 1mg/mL Pb in 2% HNO3, Thermo Scientific Chemicals
Brand: Thermo Scientific Acros 196071000
Code : 29
Additional Details : CAS Number : 7697-37-2 Weight : 0.10200kg Transport : UN number : 3264 Chem class : 8 Pack group : III
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For atomic adsorption spectroscopySpecifications
Lead standard solution | |
1 mg/ml Pb in 2% HNO3 | |
Plastic bottle | |
Brown to Red or Green | |
MFCD00011153 | |
RLJMLMKIBZAXJO-UHFFFAOYSA-N | |
λ²-lead(2+) dinitrate | |
24924 | |
331.21 | |
1.0200g/mL | |
101.0°C | |
Powder/Crystals |
997 to 1003μg/ml of Pb(2+) in 2 to 5% HNO3 (20°C) | |
10099-74-8,7697-37-2,7732-18-5 | |
100 mL | |
N2O6Pb | |
15, 5469 | |
[Pb++].[O-][N+]([O-])=O.[O-][N+]([O-])=O | |
331.20 | |
CHEBI:37187 | |
AAS | |
-3.0°C | |
1.02 |
Safety and Handling
- Lead standard solution, 1 mg/ml Pb in 2-5% HNO3
Signal Word
- Warning
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 2
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 2
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H412-Harmful to aquatic life with long lasting effects.
- H290-May be corrosive to metals.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only