missing translation for 'onlineSavingsMsg'
Learn More

Lead standard solution, for AAS, 1mg/mL Pb in 2% HNO3, Thermo Scientific Chemicals

Brand:  Thermo Scientific Acros 196075000


 View more versions of this product

Product Code. 10052460

  • £47.64 / 500mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For atomic adsorption spectroscopy
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Lead standard solution
1 mg/ml Pb in 2% HNO3
Plastic bottle
Brown to Red or Green
MFCD00011153
RLJMLMKIBZAXJO-UHFFFAOYSA-N
λ²-lead(2+) dinitrate
24924
331.21
1.0200g/mL
101.0°C
Miscible with water
997 to 1003μg/ml of Pb(2+) in 2 to 5% HNO3 (20°C)
10099-74-8, 7697-37-2, 7732-18-5
500 mL
N2O6Pb
15, 5469
[Pb++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
331.20
CHEBI:37187
AAS
-3.0°C
1.02
Powder/Crystals
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Lead standard solution, 1 mg/ml Pb in 2-5% HNO3

Signal Word
  • Warning

Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 2
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 2

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H412-Harmful to aquatic life with long lasting effects.
  • H290-May be corrosive to metals.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

RUO – Research Use Only