missing translation for 'onlineSavingsMsg'
Learn More

Lead wire, 2.0mm (0.08in) dia, Puratronic™, 99.99+% (metals basis), Thermo Scientific Chemicals

Product Code. 11350098
Click to view available options
Quantity:
50 cm
250 cm
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7439-92-1
Molecular Formula Pb
Molecular Weight (g/mol) 207.20
MDL Number MFCD00134050
InChI Key WABPQHHGFIMREM-UHFFFAOYSA-N
Synonym plumbum, metal, blei, lead, elemental, rough bullion, plomo, glover, element, flake, omaha & grant
PubChem CID 5352425
ChEBI CHEBI:27889
IUPAC Name lead
SMILES [Pb]

Specifications

Color Gray
Quantity 250 cm
Percent Purity 99.9985%
Grade Puratronic™
Odor Odorless
Weight ≈35.6g/m
Form Wire
Assay (metals basis)
Chemical Name or Material Lead wire, 2.0mm (0.08 in.) dia.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'