Learn More
Fisherbrand™ Leaf Calibration Weight Set, Class F1
Calibrate masses of class II and III weighing instruments. Fisherbrand™ Class F1 Leaf Calibration Weights are intended for use in the verification or calibration of class F2 weights and for use with weighing instruments of special accuracy.
Brand:  Fisherbrand™ FB67825
Product Code. 15545109
  • £383.00 / 1 unit
Estimated Shipment: 06-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Class F1 calibration weight
  • In compliance with OIML R111
  • Supplied individually or as a set, in a plastic case or a wooden box
Specifications
Specifications
F1
1 to 500 mg
1 mg to 500 mg