Learn More
Honeywell™ Leather Professional Footwear with Polyurethane Sole
Brand: Honeywell™ 6244411-43
This item is not returnable.
View return policy
Description
Specifications
89/686/EEC, Category II EC, EN ISO 20345:2007, ISO 9001 / 2000 | |
Leather | |
Dual density Polyurethane, Very good abrasion resistance | |
Water resistant | |
Safety Shoe |
Polyurethane | |
43 | |
Polyurethane | |
Yes |