missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LEC™ Chest Feezer
Available in three sizes, and ideal for use in all environments requiring low temperature storage
Brand: LEC Refrigeration CLT296
This item is not returnable.
View return policy
Description
Available in three sizes, and ideal for use in all environments requiring low temperature storage.- Store contents down to -45°C
- Fitted lock as standard
- Roller castors for ease of movement
- Internal and external temperature alarm
Specifications
Freezer | |
-10°C to -45°C | |
Roller | |
1260 mm | |
80 kg |
296 L | |
Chest | |
885 mm | |
50/60 Hz |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
LEC™ Chest Feezer > Capacity: 296L
Spot an opportunity for improvement?Share a Content Correction