Learn More
Glen Dimplex™ LEC™ Countertop Fridge Freezer
Save energy and reduce costs with these refrigerators. Glen Dimplex™ LEC™ Countertop Laboratory Freezers feature energy efficient R600a refrigerant on most models, heavy duty wire shelves for optimal air circulation and anti-microbial handles to help prevent growth of bacteria minimize cross-contamination.
Brand: Glen Dimplex™ LSC119UK
This item is not returnable.
View return policy
Description
- Spark free on all models
- Control panel lock to avoid accidental changes to desired settings
- Open door sensors to warn if the door has been left open (on selected models)
- Storage flexibility with adjustable shelf heights to maximize the storage potential
- Roller castors on all under-counter and freestanding models making the units easy to position