missing translation for 'onlineSavingsMsg'
Learn More

Lecithin, 90%, soybean

Product Code. 15424229
Click to view available options
Quantity:
50 g
100 g
250 g
Unit Size:
100g
250g
50g
This item is not returnable. View return policy

Product Code. 15424229

Brand: Thermo Scientific Alfa Aesar J61675.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 8002-43-5 | C42H80NO8P | 758.075 g/mol

Lecithin is used as a dietary supplement, food ingredient, and food additive. It is also used as a liquid carrier for plastics additives, dyes or pigments. Its primary functions are as an emulsifier, antioxidant, and release agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Lecithin is used as a dietary supplement, food ingredient, and food additive. It is also used as a liquid carrier for plastics additives, dyes or pigments. Its primary functions are as an emulsifier, antioxidant, and release agent.

Solubility
Partly soluble in water, partly soluble in acetone, DMSO.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: -20°C. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 8002-43-5
Molecular Formula C42H80NO8P
Molecular Weight (g/mol) 758.075
MDL Number MFCD00147406
InChI Key JLPULHDHAOZNQI-JLOPVYAASA-N
Synonym 3-sn-phosphatidylcholine, 1-hexadecanoyl-2-9e,12e-octadecadienoyl-sn-glycero-3-phosphocholine, 1,2-diacyl-sn-glycero-3-phosphocholine, 3,5,8-trioxa-4-phosphahexacosa-17,20-dien-1-aminium, 4-hydroxy-n,n,n-trimethyl-9-oxo-7-1-oxohexadecyl oxy methyl-, inner salt, 4-oxide, r, l-, a-lecithin, 2-linoleoyl-1-palmitoyl-sn-glycero-3-phosphocholine, a-phosphatidylcholine solution, l-alpha-phosphatidylcholine solution, 2-linoleoyl-1-palmitoyl-sn-glyc-ero-3-phosphocholine
PubChem CID 16213884
ChEBI CHEBI:86658
IUPAC Name [(2R)-3-hexadecanoyloxy-2-[(9E,12E)-octadeca-9,12-dienoyl]oxypropyl] 2-(trimethylazaniumyl)ethyl phosphate
SMILES CCCCCCCCCCCCCCCC(=O)OCC(COP(=O)([O-])OCC[N+](C)(C)C)OC(=O)CCCCCCCC=CCC=CCCCCC

Specifications

Color Brown to Yellow
Odor Odorless
Quantity 50 g
Beilstein 5209585
Merck Index 14,5428
Solubility Information Partly soluble in water,partly soluble in acetone,DMSO.
Percent Purity 90%
Physical Form Solid
Chemical Name or Material Lecithin, soybean

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.