Learn More
BINDER™ LED Light Bars
Product Code. 15705663 Shop All BINDER Products
Click to view available options
Length (Metric):
30 cm
50 cm
90 cm
Unit Size:
Each
Product Code. 15705663
Brand: BINDER™ 80121718
Only null left
Add to basket
Add to basket
LED light bars - Expansion set consisting of 2 pieces, attachment material: clips. For expanding the basic set of light bars
  • Expansion set consisting of 2 pieces
  • Attachment material: clips
  • For expanding the basic set of light bars
Specifications
Description LED light bars - Expansion set 900, length 90 cm, Expansion set consisting of 2 Pc., attachment material: clips. For expanding the basic set of light bars
For Use With (Equipment) BINDER Chambers KB 400, KB 720, KB ECO720, KB ECO 1020, KBF-S 720, KBF-S 1020, KBF-S ECO 720, KBF-S ECO 1020, KBF 720, KBF 1020, , KMF 720
Material Plastic
Type Lighting
For Use With (Application) Expansion of Basic Set of Light Bars
Includes 2 Pc.
Length (Metric) 90 cm
Electrical Requirements 100/240 V, 50/60 Hz
Voltage 100/240 V
Frequency 50/60 Hz