missing translation for 'onlineSavingsMsg'
Learn More

SCHOTT AG Lighting and Imaging™ LED Transmitted Light Stage for VisiLED Series

The LED Transmitted Light Stage for VisiLED Series offers professional incident and darkfield illumination. The VisiLED Backlights offer an extreme homogeneous and shadow free illumination.

Brand:  SCHOTT AG Lighting and Imaging™

Code : A0

Additional Details : Weight : 0.21000kg

 View more versions of this product

Product Code. 12826543

  • £499.08 / Each
Estimated Shipment: 27-09-2024
to see stock.
Add to basket

Description

Description

  • Professional incident and darkfield illumination.
  • The VisiLED Backlights offer an extreme homogeneous and shadow free illumination in a robust metal housing and well designed heat sink to make a maximum light density up to 20,000 cd/m2 and a long lifetime of at least 50,000 hours.
  • The VisiLED Brightfield Backlight TLS-BF features ascratch resistant SCHOTT Opalika™ surface.
  • The VisiLED Darkfield Backlight TLS-DF offers the possibility of transmitted darkfield illumination.
Specifications

Specifications

Transmitted Light Stage
3.3 in.
50 mm
VisiLED Controller MC750, MC1100 or MC1500
15 mm
Transmitted Light Stage TLS-BF Brightfield, Luminance: 20 000 cd/m2 (in center)
83.9 mm
Stereo Microscopy Illumination
0.6 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers