Learn More
BINDER™ Left Access Port (100 mm)
For Use With: KBF P 720, KBF P 240
Brand: BINDER™ 8012-1341
This item is not returnable.
View return policy
Description
- For use on left
- Includes silicone plug
- For use with a wide range of BINDER™ incubators and chambers
Specifications
Access port with silicone plug - 100 mm, left | |
Silicone | |
Silicone plug |
BINDER Chambers Series KBF P | |
Access Port | |
100 mm |