Learn More
Thermo Scientific™ Legionella Acid Buffer Solution
Conveniently prepare water samples when isolating Legionella species according to ISO 11731 water quality testing specifications.
Brand: Thermo Scientific™ GFB01
Additional Details : CAS Number : 7647-01-0 Weight : 6.00000kg
Description
Conveniently prepare water samples when isolating and enumerating Legionella species according to DIN EN ISO 11731 water quality testing specifications using Thermo Scientific™ Legionella Acid Buffer Solution.
- Ready-to-use: save time spent preparing your own buffer solution
- Sterile: additional sterilization by autoclaving is unnecessary
- Easy to transport and store: high-quality, transparent PET bottle has a leak-proof screw top made of HDPE and is lighter and more shatter-resistant than glass, making it better for handling in the laboratory as well as shipping
- Ergonomic design: the bottle has a wide drip edge that makes pouring easier and prevents dripping
- Easy measurement: 50mL gradations allow for easy measurement of the remaining content
Specifications
Colorless, clear | |
Bottle | |
Buffer Solution | |
1000mL | |
6/Pk. |
Legionella Acid Buffer Solution | |
Germany | |
1 L | |
Sterile, tamper-proof seal |