missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Legionella BCYE Medium with Antibiotics

Product Code. 15888252
Click to view available options
Fill Volume:
17 mL
Fill Weight:
17g
Unit Size:
Pack of 10
This item is not returnable. View return policy

Product Code. 15888252

Brand: Thermo Scientific™ PO5325A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Isolate Legionellaceae in environmental samples with this selective medium which conforms to ISO standards and yields optimal recovery of Legionellaceae.

Isolate Legionella from water samples using Thermo Scientific™ Legionella BCYE Medium with Antibiotics, a selective medium that conforms to ISO11731:2016 Water quality – Enumeration of Legionella and exhibits optimal recovery of Legionellaceae in short incubation periods from environmental samples.

Legionella are aerobic, motile, nutritionally fastidious, pleomorphic, gram-negative rods that require special growth factors in culture media to grow. Legionella BCYE Medium with Antibiotics is a recommended medium for the isolation of Legionella from water samples:

  • Yields optimal recovery in a short incubation period
  • Conforms to ISO11731:2016 standards
  • Quality control tested according to ISO11133:2014, including membrane filtration method
  • Selective against non-target bacteria like Enterococcus faecalis and Escherichia coli
  • Easy-to-read results as Legionella form gray-blue colonies

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific™ brand.

TRUSTED_SUSTAINABILITY

Specifications

Color Traffic black to Jet black, opaque
Content And Storage 2°C to 12°C
Description Legionella BCYE Medium with Antibiotics
Format 90mm Monoplate
Manufacturing Origin Germany
Product Type Agar
Fill Volume 17 mL
Fill Weight 17g
Packaging Type Flow Wrapped - Single Layer
Quantity 10/Pk.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.

Mange tak! Din feedback er blevet sendt. Hos Fisher Scientific arbejder vi altid på at forbedre vores indhold for dig. Vi sætter pris på din feedback.