missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Legionella BMPA Selective Agar
Isolate Legionella from clinical specimens with a selective microbiological medium which yields optimal recovery of Legionellaceae in short incubation periods.
Brand: Thermo Scientific™ PO0324A
This item is not returnable.
View return policy
Description
Isolate Legionella from clinical specimens using Thermo Scientific™ Legionella BMPA Selective Agar, a selective medium that exhibits optimal recovery of Legionellaceae in short incubation periods from both environmental samples and clinical specimens.
Legionella are aerobic, motile, nutritionally fastidious, pleomorphic, gram-negative rods that require special growth factors in culture media to grow. Transmission occurs through aerosolization or aspiration of contaminated water, not person-to-person. Legionellosis refers to two distinct clinical syndromes: Legionnaires’ disease, which most often presents as severe pneumonia, and Pontiac fever, which is an acute, febrile, self-limiting, viral-like illness. Over forty species of Legionella have been characterized; however, Legionella pneumophila remains the most prevalent etiological agent of Legionnaires’ disease. Legionella can be isolated from respiratory tract secretions of all types, sterile body fluids such as blood, and biopsy material.This product may not be available in all countries or territories. Please consult your local representative for availability.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
Black, opaque | |
90mm Monoplate | |
Legionella BMPA Selective Agar | |
19.5g ± 1.0g | |
10/Pk. |
2°C to 10°C | |
United Kingdom | |
Agar | |
Flow Wrapped - Single Layer |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction