missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Legionella Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA17227
This item is not returnable.
View return policy
Description
PA1-7227 detects Legionella pneumophila. PA1-7227 has been successfully used in immunofluorescence procedures. The PA1-7227 immunogen is a whole cell preparation of Legionella pneumophila; ATCC #33152. Recommended short term storage of less than 6 months at 2-8oC. For extended storage, aliquot and freeze.
Legionella is found in aquatic environments where its growth is supported by amoebae. In human infections, Legionella acts as an intracellular parasite and multiplies in alveolar macrophages. The genus Legionella contains 39 recognized species. The immunological diversity of Legionella is due to the existence of a variety of serogroups. Legionella pneumophila has the most with a total of 14 serogroups.Specifications
Legionella | |
Polyclonal | |
Unconjugated | |
L. pneumophila; Legionellosis | |
A whole cell preparation of Legionella pneumophila; ATCC #33152. | |
1 mL | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
4-5 mg/mL | |
PBS with 0.1% sodium azide; pH 7.2 | |
Rabbit | |
IgG fraction | |
RUO | |
Bacteria | |
Antibody | |
IgG |