Learn More
Gibco™ Leibovitz's L-15 Medium, GlutaMAX™ Supplement

Description
Leibovitz's L-15 Medium supports the growth of HEP-2 monkey kidney cells and primary explants of embryonic and adult human tissue. L-15 is buffered by phosphates and free base amino acids instead of sodium bicarbonate. This medium is designed for supporting cell growth in environments without CO2 equilibration.
This L-15 is modified as follows:
With: Galactose, Phenol Red, GlutaMAX™, Sodium Pyruvate
Without: Glucose, HEPES, Sodium Bicarbonate
The complete formulation is available.
Gibco™ L-15 with GlutaMAX™ supplement minimizes toxic ammonia build-up and improves cell viability and growth in an easy-to-use format.
L-15 contains no proteins, lipids, or growth factors. Therefore, L-15 requires supplementation, commonly with 10% Fetal Bovine Serum (FBS).
Specifications
Specifications
| Classification | Animal Origin-free |
| Concentration | 1 X |
| Form | Liquid |
| Product Type | Leibovitz's L-15 Medium |
| Sterility | Sterile-filtered |
| With Additives | GlutaMAX, Phenol Red, Sodium Pyruvate, Galactose |
| Without Additives | No HEPES, No Sodium Bicarbonate, No Glucose |
| Manufacturing Quality | cGMP-compliant under the ISO 13485 standard |
| Product Line | Gibco, GlutaMAX |
| Quantity | 500 mL |
| Show More |
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.