Learn More
Tocris™ Lenalidomide 4'-PEG2-amine
Cereblon ligand with PEG linker and terminal amine for onward chemistry
Brand: Tocris™ 7093/25
Additional Details : Weight : 0.00970kg
Description
Lenalidomide 4'-PEG2-amine is a functionalized cereblon ligand for PROTAC™ research and development; incorporates an E3 ligase ligand plus a PEG2 linker with terminal amine ready for conjugation to a target protein ligand. Part of a range of functionalized tool molecules for PROTAC R&D. PROTAC™ is a registered trademark of Arvinas Operations, Inc., and is used under license.Specifications
3-[4-[[2-[2-(2-(2-Aminoethoxy)ethoxy]ethyl]amino]-1,3-dihydro-1-oxo-2 H-isoindol-2-yl]-2,6-piperidinedione dihydrochloride | |
Degrader Building Blocks | |
0.95 |
25 mg | |
C19H26N4O50.2 HCl |