missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisher BioReagents™ Lennox Luria-Bertani Broth
Used for the culturing of E. coli.
£127.20 - £403.86
Specifications
Media Type | Lennox Luria-Bertani Broth |
---|---|
Product Type | Microbiology Media |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
12881650
|
Fisher BioReagents™
12881650 |
500 g |
£127.20
500g |
In Stock
Log in to see stock. |
|||||
12871650
|
Fisher BioReagents™
12871650 |
2000 g |
£403.86
2kg |
Estimated Shipment: 09-10-2024
Log in to see stock. |
|||||
Description
- Large granules
Specifications
Lennox Luria-Bertani Broth | |
Microbiology Media |